missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine disodium salt dihydrate

Catalog No. AAJ6177022
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6177022 100 g
AAJ6177036 500 g
AAJ61770A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6177022 Supplier Thermo Scientific Chemicals Supplier No. J6177022

CAS: 122666-87-9 | C9H13NNa2O5 | 261.19 g/mol

Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 122666-87-9
Molecular Formula C9H13NNa2O5
Molecular Weight (g/mol) 261.19
MDL Number MFCD03840558
InChI Key WFTCFVUQORELJZ-CMIWSEPMNA-L
Synonym tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate
PubChem CID 71587456
SMILES O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Color Brown to White
Solubility Information Soluble in water.
IUPAC Name 2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium
Quantity 100 g
Formula Weight 261.18 (225.15 Anhydrous)
Optical Rotation −13° (c=4 in Water)
Sensitivity Very Hygroscopic
Physical Form Powder
Chemical Name or Material L-Tyrosine disodium salt dihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only