Learn More
Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent
CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol
Supplier: Thermo Scientific Chemicals J6351122
| Quantity | 100 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C9H11NO3 | |
| MFCD00002606 | |
| l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
| CHEBI:17895 |
Specifications
| >300°C | |
| 60-18-4 | |
| C9H11NO3 | |
| 392441 | |
| l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
| N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
| 100 g | |
| 6057 | |
| 181.19 | |
| Cell Culture Reagent |
| L-Tyrosine | |
| Brown to White | |
| MFCD00002606 | |
| 14,9839 | |
| OUYCCCASQSFEME-SVGMAFHSNA-N | |
| (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 181.19 | |
| CHEBI:17895 | |
| −11° (c=4 in 1N HCl) | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-460-4
RTECSNumber : YP2275600
TSCA : Yes
Recommended Storage : Ambient temperatures