missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. p-7137219
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24435G 5 g
T244325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24435G Supplier TCI America Supplier No. T24435G

Chemical Identifiers

CAS 53587-11-4
Molecular Formula C23H25NO6S
Molecular Weight (g/mol) 443.51
MDL Number MFCD00038959
InChI Key PJGVHBLZZQDFFM-RSAXXLAASA-N
Synonym h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
PubChem CID 104540
IUPAC Name 4-methylbenzene-1-sulfonic acid; benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 177°C
Color White
Quantity 5 g
Formula Weight 443.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine Benzyl Ester p-Toluenesulfonate