missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, 98.5-101.5%, Spectrum™ Chemical

$305.36 - $5,208.04

Chemical Identifiers

CAS 60-18-4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611389
View Documents Promotion Details
N/A 100 g Poly Pail
Each for $305.36
Only null left
 
18611388
View Documents Promotion Details
N/A 1 kg Poly Pail
Each for $1,772.71
Only null left
 
18611390
View Documents Promotion Details
N/A 5 kg Poly Pail
Each for $5,208.04
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60-18-4

Specifications

60-18-4
30 ppm
-10.0° to -12.3°
Reagent
1
0.003
98.5 to 101.5%
L-Tyrosine
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.