missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7136891
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16575G 5 g
T165725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16575G Supplier TCI America Supplier No. T16575G
Only null left

Chemical Identifiers

CAS 7524-52-9
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00066134
InChI Key XNFNGGQRDXFYMM-PPHPATTJSA-N
Synonym l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
PubChem CID 2734891
IUPAC Name methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 220°C
Quantity 5 g
Formula Weight 254.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophan Methyl Ester Hydrochloride