missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophan Ethyl Ester Hydrochloride 95.0+%, TCI America™
$35.41 - $35.41
Chemical Identifiers
| CAS | 2899-28-7 |
|---|---|
| Molecular Formula | C13H17ClN2O2 |
| Molecular Weight (g/mol) | 268.74 |
| MDL Number | MFCD00038993 |
| InChI Key | PESYCVVSLYSXAK-UHFFFAOYNA-N |
| Synonym | l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920 |
| PubChem CID | 134519 |
| IUPAC Name | hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride |
| SMILES | [H+].[Cl-].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12 |
Chemical Identifiers
| 2899-28-7 | |
| 268.74 | |
| PESYCVVSLYSXAK-UHFFFAOYNA-N | |
| 134519 | |
| [H+].[Cl-].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12 |
| C13H17ClN2O2 | |
| MFCD00038993 | |
| l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920 | |
| hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride |
Specifications
| 222°C | |
| C13H17ClN2O2 | |
| l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920 | |
| [H+].[Cl-].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12 | |
| 5 g | |
| 134519 | |
| ≥95.0% (HPLC,T) | |
| L-Tryptophan Ethyl Ester Hydrochloride |
| 2899-28-7 | |
| MFCD00038993 | |
| PESYCVVSLYSXAK-UHFFFAOYNA-N | |
| hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride | |
| 268.74 | |
| 268.74 | |
| Crystalline Powder |
Safety and Handling
TSCA : No