missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Tryptophan, Calbiochem™,

Catalog No. 6540100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6540100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6540100GM Supplier MilliporeSigma™ Supplier No. 6540100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Essential amino acid for human development that serves as a precursor of serotonin.

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White
Packaging Glass Bottle
Quantity 100 g
Percent Purity ≥98.5% (Titration)
Grade Biochemical
Physical Form Crystalline Solid
Chemical Name or Material L-Tryptophan