missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, ≥99%, MP Biomedicals™

Catalog No. ICN10315191
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10315191 1 kg
ICN10315125 25 g
ICN10315180 100 g
ICN10315190 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10315191 Supplier MP Biomedicals Inc Supplier No. 0210315191

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Tryptophan is an amino acid precursor of serotonin and melatonin

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 1 kg
Formula Weight 204.22
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophan
EINECSNumber 200-795-6
Recommended Storage Store at Room temperature (15 to 30°C).