missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tryptophan, 99%

CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol

Supplier:  Thermo Scientific Chemicals A1023022

 View more versions of this product

Catalog No. AAA1023022


Only null left

Description

Description

L-tryptophan is an essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. It acts as a natural dietary supplement and used as an antidepressant, anxiolytic and sleep aid. It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-tryptophan is an essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. It acts as a natural dietary supplement and used as an antidepressant, anxiolytic and sleep aid. It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.

Solubility
Soluble in water, ammonia, hot alcohol, alkali hydroxides and dilute acids. Slightly soluble in acetic acid and ethanol. Insoluble in chloroform and ethyl ether.

Notes
Incompatible with strong acids and strong oxidizing agents.
Specifications

Specifications

280°C 285°C
73-22-3
C11H12N2O2
86197
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
QIVBCDIJIAJPQS-VIFPVBQESA-N
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
204.23
CHEBI:16828
204.23
L-Tryptophan
99%
MFCD00064340
14,9797
Soluble in water,ammonia,hot alcohol,alkali hydroxides and dilute acids. Slightly soluble in acetic acid and ethanol. Insoluble in chloroform and ethyl ether.
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
100 g
6305
−31° (c=1 in water)
99%

Chemical Identifiers

73-22-3
204.23
QIVBCDIJIAJPQS-VIFPVBQESA-N
6305
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C11H12N2O2
MFCD00064340
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
CHEBI:16828
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-795-6

RTECSNumber : YN6130000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only