missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Tryptophan, 99%

L(-)-Tryptophan, CAS # 73-22-3, is an essential amino acid necessary for the biosynthesis of proteins. | CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol

Supplier:  Thermo Scientific Chemicals 140591000

Quantity 100 g
Packaging Plastic Bottle

 View more versions of this product

Catalog No. AC140591000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
  • It is a biochemical precursor of niacin, indole alkaloids, and serotonin

Applications

  • L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
  • It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
Specifications

Chemical Identifiers

73-22-3
204.23
QIVBCDIJIAJPQS-VIFPVBQESA-N
6305
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C11H12N2O2
MFCD00064340
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
CHEBI:16828
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

280.0°C to 285.0°C
99%
White to White-Yellow
0.5% max. (1 g, 105°C)
Plastic Bottle
MFCD00064340
22, 546
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
QIVBCDIJIAJPQS-VIFPVBQESA-N
− 31.10 (20.00°C c=1,H20)
204.23
6305
204.23
Crystalline Powder
L(-)-Tryptophan
73-22-3
Authentic
99%
C11H12N2O2
− 31.10
15, 9977
Solubility in water: 11.4g/L (25°C). Other solubilities: soluble in alkali hydroxides and hot alcohol, insoluble in chloroform
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
100 g
CHEBI:16828
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-795-6

RTECSNumber : YN6130000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions