missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Thyroxine Sodium Salt Pentahydrate 98.0+%, TCI America™
$52.19 - $149.13
Chemical Identifiers
| CAS | 55-03-8 |
|---|---|
| Molecular Formula | C15H10I4NNaO4 |
| Molecular Weight (g/mol) | 798.856 |
| MDL Number | MFCD00002596 |
| InChI Key | YDTFRJLNMPSCFM-YDALLXLXSA-M |
| Synonym | (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic Acid Sodium Salt, Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate |
| PubChem CID | 23666112 |
| ChEBI | CHEBI:6446 |
| IUPAC Name | sodium;(2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate |
| SMILES | C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O)I)I)CC(C(=O)[O-])N.[Na+] |
Chemical Identifiers
| 55-03-8 | |
| 798.856 | |
| YDTFRJLNMPSCFM-YDALLXLXSA-M | |
| 23666112 | |
| sodium;(2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate |
| C15H10I4NNaO4 | |
| MFCD00002596 | |
| (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic Acid Sodium Salt, Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate | |
| CHEBI:6446 | |
| C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O)I)I)CC(C(=O)[O-])N.[Na+] |
Specifications
| 55-03-8 | |
| MFCD00002596 | |
| 2811 | |
| YDTFRJLNMPSCFM-YDALLXLXSA-M | |
| sodium;(2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate | |
| 23666112 | |
| 798.86 | |
| Crystalline Powder |
| C15H10I4NNaO4 | |
| 100 mg | |
| (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic Acid Sodium Salt, Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate | |
| C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O)I)I)CC(C(=O)[O-])N.[Na+] | |
| 798.856 | |
| CHEBI:6446 | |
| ≥98.0% (T) | |
| L-Thyroxine Sodium Salt Pentahydrate |
Safety and Handling
RTECSNumber : XP3583000
TSCA : Yes