missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Thymidine

Catalog No. AC464550010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464550010 1 g
AC464550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464550010 Supplier Thermo Scientific Chemicals Supplier No. 464550010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Format Crystalline Powder
Molecular Weight (g/mol) 242.23
Product Type L-Thymidine
Solution Clarity (1% aq. soln.) Clear colorless solution
Source synthetic source
Content And Storage Refrigerator +4°C
Shipping Condition normal conditions
Purity ≥98% by HPLC
Quantity 1 g
Color White to almost white
ShelfLife 5 years

RUO – Research Use Only