missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Threonine 99.0+%, TCI America™

Catalog No. p-7136157
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
T023025G 25 g
T0230100G 100 g
T0230500G 500 g
3 options

Catalog No. T023025G

Supplier: TCI America T023025G

Only null left

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 119.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Threonine
EINECSNumber (9)-1584
RTECSNumber XO8590000
TSCA Yes