missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine Benzyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. T26775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26775G 5 g
T26771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26775G Supplier TCI America Supplier No. T26775G

Chemical Identifiers

CAS 33645-24-8
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.703
InChI Key IDZGTFSDZJVMSD-SCYNACPDSA-N
Synonym H-Thr-OBzl.HCl
PubChem CID 16216248
IUPAC Name benzyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride
SMILES CC(C(C(=O)OCC1=CC=CC=C1)N)O.Cl

Specifications

Melting Point 130°C
Color White
Quantity 5 g
Formula Weight 245.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine Benzyl Ester Hydrochloride
TSCA No
Recommended Storage Refrigerator