missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine Allo-free base, 990%, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Assay Percent Range | 99% |
| Quantity | 10 g |
| Formula Weight | 119.1 |
| Percent Purity | 99.5% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Threonine, Allo Free |
Sécurité et manipulation
| EINECSNumber | 200-774-1 |
| Recommended Storage | Store at Room temperature (15 to 30°C). |