missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, 98+%

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

Supplier:  Thermo Scientific Chemicals A1685122

 View more versions of this product

Catalog No. AAA1685122


Only null left

Description

Description

L-Threonine is used to alleviate anxiety and mild depression. It is an important component of many proteins such as tooth enamel, collagen, and elastin. It is an important amino acid in the nervous system and plays a vital role in porphyrin and fat metabolism. It is useful in intestinal disorders and indigestion. It is also used as a prodrug to reliably elevate brain glycine levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Threonine is used to alleviate anxiety and mild depression. It is an important component of many proteins such as tooth enamel, collagen, and elastin. It is an important amino acid in the nervous system and plays a vital role in porphyrin and fat metabolism. It is useful in intestinal disorders and indigestion. It is also used as a prodrug to reliably elevate brain glycine levels.

Solubility
Soluble in water. Insoluble in ethanol, ethyl ether and chloroform.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

255°C (decomposition)
72-19-5
MFCD00064270
14,9380
Soluble in water. Insoluble in ethanol,ethyl ether and chloroform.
CC(O)C(N)C(O)=O
100 g
6288
−28° (c=5 in Water)
≥98%
L-Threonine
C4H9NO3
1721646
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
AYFVYJQAPQTCCC-UHFFFAOYNA-N
2-amino-3-hydroxybutanoic acid
119.12
CHEBI:16857
119.12

Chemical Identifiers

72-19-5
119.12
AYFVYJQAPQTCCC-UHFFFAOYNA-N
6288
CC(O)C(N)C(O)=O
C4H9NO3
MFCD00064270
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
CHEBI:16857
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-774-1

RTECSNumber : XO8590000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only