missing translation for 'onlineSavingsMsg'
Learn More

L-Thioproline 98.0+%, TCI America™

Catalog No. p-7136150
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T021925G 25 g
T0219500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T021925G Supplier TCI America Supplier No. T021925G

Chemical Identifiers

CAS 34592-47-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00005212
InChI Key DZLNHFMRPBPULJ-VKHMYHEASA-N
Synonym l-thioproline, r-thiazolidine-4-carboxylic acid, l-thiaproline, l-thiazolidine-4-carboxylic acid, 4r-1,3-thiazolidine-4-carboxylic acid, h-thz-oh, r-4-thiazolidinecarboxylic acid, 4-thiaproline, thioproline, gamma-thioproline
PubChem CID 93176
ChEBI CHEBI:45171
IUPAC Name (4R)-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NCS1)C(=O)O

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 133.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Thioproline
EINECSNumber (9)-0807
RTECSNumber XJ5425500
TSCA No