missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Thiazolidine-4-carboxylic acid, 98%

Catalog No. p-199957
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138640100 10 g Glass bottle
AC138640250 25 g Glass bottle
AC138641000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138640100 Supplier Thermo Scientific Chemicals Supplier No. 138640100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34592-47-7 | C4H7NO2S | 133.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34592-47-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.16
MDL Number MFCD00005212
InChI Key DZLNHFMRPBPULJ-VKHMYHEASA-N
Synonym l-thioproline, r-thiazolidine-4-carboxylic acid, l-thiaproline, l-thiazolidine-4-carboxylic acid, 4r-1,3-thiazolidine-4-carboxylic acid, h-thz-oh, r-4-thiazolidinecarboxylic acid, 4-thiaproline, thioproline, gamma-thioproline
PubChem CID 93176
ChEBI CHEBI:45171
IUPAC Name (4R)-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NCS1)C(=O)O

Specifications

Melting Point 203°C
Color Pink to White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Merck Index 09, 9040
Specific Rotation Condition −101° (20°C c=1,1N HCl)
Specific Rotation -101
Solubility Information Solubility in water: 28.5g/L (20°C). Other solubilities: soluble in 1n hcl,practically insoluble in alcohol
Formula Weight 133.16
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material L(-)-Thiazolidine-4-carboxylic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 252-106-3
RTECSNumber XJ5425500

RUO – Research Use Only