missing translation for 'onlineSavingsMsg'
Learn More

L-Thiazolidine-4-carboxylic acid, 98%

Catalog No. AAA1503309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1503309 10 g
AAA1503318 50 g
AAA1503330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1503309 Supplier Thermo Scientific Chemicals Supplier No. A1503309

CAS: 34592-47-7 | C4H7NO2S | 133.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34592-47-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00005212
InChI Key DZLNHFMRPBPULJ-VKHMYHEASA-N
Synonym l-thioproline, r-thiazolidine-4-carboxylic acid, l-thiaproline, l-thiazolidine-4-carboxylic acid, 4r-1,3-thiazolidine-4-carboxylic acid, h-thz-oh, r-4-thiazolidinecarboxylic acid, 4-thiaproline, thioproline, gamma-thioproline
PubChem CID 93176
ChEBI CHEBI:45171
IUPAC Name (4R)-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NCS1)C(=O)O

Specifications

Melting Point 192°C to 200°C (decomposition)
Quantity 10 g
UN Number UN2811
Beilstein 81065
Merck Index 14,9445
Optical Rotation −141° (c=1.3 in Water)
Formula Weight 133.17
Percent Purity 98%
Chemical Name or Material L-Thiazolidine-4-carboxylic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 252-106-3
RTECSNumber XJ5425500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only