missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Theanine 98.0+%, TCI America™
$165.90 - $561.08
Chemical Identifiers
| CAS | 3081-61-6 |
|---|---|
| Molecular Formula | C7H14N2O3 |
| Molecular Weight (g/mol) | 174.20 |
| MDL Number | MFCD00059653 |
| InChI Key | DATAGRPVKZEWHA-UHFFFAOYNA-N |
| Synonym | l-theanine, theanine, theanin, s-2-amino-5-ethylamino-5-oxopentanoic acid, n-ethyl-l-glutamine, suntheanine, n5-ethyl-l-glutamine, n 5-ethyl-l-glutamine, unii-8021pr16qo |
| PubChem CID | 439378 |
| ChEBI | CHEBI:17394 |
| IUPAC Name | 2-amino-4-(ethylcarbamoyl)butanoic acid |
| SMILES | CCNC(=O)CCC(N)C(O)=O |
Chemical Identifiers
| 3081-61-6 | |
| 174.20 | |
| DATAGRPVKZEWHA-UHFFFAOYNA-N | |
| 439378 | |
| 2-amino-4-(ethylcarbamoyl)butanoic acid |
| C7H14N2O3 | |
| MFCD00059653 | |
| l-theanine, theanine, theanin, s-2-amino-5-ethylamino-5-oxopentanoic acid, n-ethyl-l-glutamine, suntheanine, n5-ethyl-l-glutamine, n 5-ethyl-l-glutamine, unii-8021pr16qo | |
| CHEBI:17394 | |
| CCNC(=O)CCC(N)C(O)=O |
Specifications
| 3081-61-6 | |
| White | |
| MFCD00059653 | |
| l-theanine, theanine, theanin, s-2-amino-5-ethylamino-5-oxopentanoic acid, n-ethyl-l-glutamine, suntheanine, n5-ethyl-l-glutamine, n 5-ethyl-l-glutamine, unii-8021pr16qo | |
| CCNC(=O)CCC(N)C(O)=O | |
| 174.20 | |
| CHEBI:17394 | |
| ≥98.0% (T) | |
| L-Theanine |
| 207°C | |
| C7H14N2O3 | |
| 1 g | |
| DATAGRPVKZEWHA-UHFFFAOYNA-N | |
| 2-amino-4-(ethylcarbamoyl)butanoic acid | |
| 439378 | |
| 174.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No