missing translation for 'onlineSavingsMsg'
Learn More

L-Theanine 98.0+%, TCI America™

Catalog No. T09541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09541G 1 g
T09545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09541G Supplier TCI America Supplier No. T09541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3081-61-6
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.20
MDL Number MFCD00059653
InChI Key DATAGRPVKZEWHA-UHFFFAOYNA-N
Synonym l-theanine, theanine, theanin, s-2-amino-5-ethylamino-5-oxopentanoic acid, n-ethyl-l-glutamine, suntheanine, n5-ethyl-l-glutamine, n 5-ethyl-l-glutamine, unii-8021pr16qo
PubChem CID 439378
ChEBI CHEBI:17394
IUPAC Name 2-amino-4-(ethylcarbamoyl)butanoic acid
SMILES CCNC(=O)CCC(N)C(O)=O

Specifications

Melting Point 207°C
Color White
Quantity 1 g
Formula Weight 174.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Theanine