missing translation for 'onlineSavingsMsg'
Learn More

L-Theanine, 98%

Catalog No. p-7077792
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459340010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459340010 Supplier Thermo Scientific Chemicals Supplier No. 459340010
Only null left

CAS: 3081-61-6 | C7H14N2O3 | 174.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3081-61-6
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.20
MDL Number MFCD00059653
InChI Key DATAGRPVKZEWHA-UHFFFAOYNA-N
Synonym l-theanine, theanine, theanin, s-2-amino-5-ethylamino-5-oxopentanoic acid, n-ethyl-l-glutamine, suntheanine, n5-ethyl-l-glutamine, n 5-ethyl-l-glutamine, unii-8021pr16qo
PubChem CID 439378
ChEBI CHEBI:17394
IUPAC Name (2S)-2-amino-5-(ethylamino)-5-oxopentanoic acid
SMILES CCNC(=O)CCC(N)C(O)=O

Specifications

Melting Point 206.0°C to 219.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3CH2NHC(=O)CH2CH2CH(NH2)C(=O)OH
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether, alcohols
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material L-Theanine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 221-379-

RUO – Research Use Only