missing translation for 'onlineSavingsMsg'
Learn More

L-Tetrahydropalmatine 98.0+%, TCI America™

Supplier:  TCI America T3311250MG

Catalog No. T3311250MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-Tetrahydropalmatine
142°C
C21H25NO4
250 mg
AEQDJSLRWYMAQI-KRWDZBQOSA-N
(12bS)-3,4,10,11-tetramethoxy-7,8,12b,13-tetrahydro-5H-6-azatetraphene
72301
355.43
Crystalline Powder
483-14-7
White-Yellow
MFCD03265591
tetrahydropalmatine, l-tetrahydropalmatine, rotundine, --tetrahydropalmatine, s-tetrahydropalmatine, caseanine, gindarine, hyndarine, --corydalis, unii-3x69co5i79
COC1=CC2=C(C=C1OC)[C@@H]1CC3=C(CN1CC2)C(OC)=C(OC)C=C3
355.43
CHEBI:16563
≥98.0% (HPLC)

Chemical Identifiers

483-14-7
355.43
AEQDJSLRWYMAQI-KRWDZBQOSA-N
72301
(12bS)-3,4,10,11-tetramethoxy-7,8,12b,13-tetrahydro-5H-6-azatetraphene
C21H25NO4
MFCD03265591
tetrahydropalmatine, l-tetrahydropalmatine, rotundine, --tetrahydropalmatine, s-tetrahydropalmatine, caseanine, gindarine, hyndarine, --corydalis, unii-3x69co5i79
CHEBI:16563
COC1=CC2=C(C=C1OC)[C@@H]1CC3=C(CN1CC2)C(OC)=C(OC)C=C3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : HQ1792500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions