missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine Methylamide 96.0+%, TCI America™

Catalog No. p-7131856
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01681G 1 g
L01685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01681G Supplier TCI America Supplier No. L01681G
Only null left

Chemical Identifiers

CAS 89226-12-0
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.218
MDL Number MFCD00673329
InChI Key BPKJNEIOHOEWLO-RXMQYKEDSA-N
Synonym L-tert-Leucylaminomethane
PubChem CID 445856
IUPAC Name (2S)-2-amino-N,3,3-trimethylbutanamide
SMILES CC(C)(C)C(C(=O)NC)N

Specifications

Color White
Quantity 1 g
Formula Weight 144.22
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-tert-Leucine Methylamide