missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-tert-Leucine Methylamide 96.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 144.22 |
| Percent Purity | ≥96.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-tert-Leucine Methylamide |
Safety and Handling
| TSCA | No |