missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. L01881G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01881G 1 g
L01885G 5 g
L018825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L01881G Supplier TCI America Supplier No. L01881G
Only null left

Chemical Identifiers

CAS 63038-27-7
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00040604
InChI Key HRTQWUHFSXVRPY-XZNNNFJINA-N
Synonym l-tert-leucine methyl ester hydrochloride, h-tle-ome.hcl, s-methyl 2-amino-3,3-dimethylbutanoate hydrochloride, methyl 2s-2-amino-3,3-dimethylbutanoate hydrochloride, s-2-amino-3,3-dimethylbutyric acid methyl ester hydrochloride, methyl 3-methyl-l-valinate hydrochloride, pubchem11076, methyl l-tert-leucinate hydrochloride, l-tert-leucine methyl ester hcl, l-tert-leucine methyl-ester hydrochloride
PubChem CID 16217630
IUPAC Name methyl (2S)-2-amino-3,3-dimethylbutanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)C(C)(C)C

Specifications

Color White
Quantity 1 g
Formula Weight 181.66
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-tert-Leucine Methyl Ester Hydrochloride