missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid diammonium salt, 99%

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

$132.16 - $132.16

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211812500
SDS View Documents
Thermo Scientific Chemicals
211812500
250 g Glass bottle
Each for $132.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3164-29-2
184.15
NGPGDYLVALNKEG-OLXYHTOASA-N
13652278
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
C4H12N2O6
MFCD00013073
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
CHEBI:63075

Specifications

3164-29-2
1.6000g/mL
Authentic
Glass bottle
[-CH(OH)CO2NH4]2
250 g
+ 32.40 (20.00°C c=1.84,H2O)
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
184.15
CHEBI:63075
≥98.5%
Crystals
White
6.0 to 7.0 (5% soln.)
0.99
C4H12N2O6
MFCD00013073
09, 591
+31.4° to +33.4° (20°C, 589nm) (c=1.84, H2O)
NGPGDYLVALNKEG-OLXYHTOASA-N
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
13652278
184.15
0.005% max.
L(+)-Tartaric acid diammonium salt, 0.99
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-618-9

RTECSNumber : WW8050000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only