missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid diammonium salt, 99%, Thermo Scientific Chemicals

Catalog No. AC211812500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211812500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC211812500 Supplier Thermo Scientific Chemicals Supplier No. 211812500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifications

Color White
Density 1.6000g/mL
pH 6.0 to 7.0 (5% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula [-CH(OH)CO2NH4]2
Quantity 250 g
Merck Index 09, 591
Specific Rotation Condition + 32.40 (20.00°C c=1.84,H2O)
Specific Rotation +31.4° to +33.4° (20°C, 589nm) (c=1.84, H2O)
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid diamine
Formula Weight 184.15
Percent Purity ≥98.5%
Insoluble Matter 0.005% max.
Physical Form Crystals
Chemical Name or Material L(+)-Tartaric acid diammonium salt, 0.99
Show More Show Less
EINECSNumber 221-618-9
RTECSNumber WW8050000
TSCA TSCA

RUO – Research Use Only