missing translation for 'onlineSavingsMsg'
Learn More

L-Serine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118395
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02675G 5 g
B026725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02675G Supplier TCI America Supplier No. B02675G

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.578
MDL Number MFCD00063680
InChI Key NDBQJIBNNUJNHA-DFWYDOINSA-N
Synonym l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
PubChem CID 2723730
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point 161°C
Color White
Quantity 5 g
Formula Weight 155.58
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Serine Methyl Ester Hydrochloride
EINECSNumber (2)-4026
TSCA No