missing translation for 'onlineSavingsMsg'
Learn More

L-Serine Methyl Ester Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America B026725G

Quantity 25 g

 View more versions of this product

Catalog No. B026725G


Only null left

Description

Description

Specifications

Chemical Identifiers

5680-80-8
155.578
NDBQJIBNNUJNHA-DFWYDOINSA-N
2723730
COC(=O)C(CO)N.Cl
C4H10ClNO3
MFCD00063680
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride

Specifications

161°C
5680-80-8
C4H10ClNO3
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
COC(=O)C(CO)N.Cl
25 g
2723730
≥98.0% (T)
L-Serine Methyl Ester Hydrochloride
White
MFCD00063680
NDBQJIBNNUJNHA-DFWYDOINSA-N
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
155.578
155.58
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4026

TSCA : No

SDS
Documents

Documents

Promotions

Promotions