missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Serinamide hydrochloride, 98%, Thermo Scientific™
$112.38 - $112.38
Chemical Identifiers
| CAS | 65414-74-6 |
|---|---|
| Molecular Formula | C3H9ClN2O2 |
| Molecular Weight (g/mol) | 140.567 |
| MDL Number | MFCD00067560 |
| InChI Key | VURWDDZIWBGXCK-DKWTVANSSA-N |
| Synonym | l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl |
| PubChem CID | 16212403 |
| IUPAC Name | (2S)-2-amino-3-hydroxypropanamide;hydrochloride |
| SMILES | C(C(C(=O)N)N)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC303160010
|
Thermo Scientific Chemicals
303160010 |
1 g | Glass Bottle |
Each for $112.38
|
|
||||
Chemical Identifiers
| 65414-74-6 | |
| 140.567 | |
| VURWDDZIWBGXCK-DKWTVANSSA-N | |
| 16212403 | |
| C(C(C(=O)N)N)O.Cl |
| C3H9ClN2O2 | |
| MFCD00067560 | |
| l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl | |
| (2S)-2-amino-3-hydroxypropanamide;hydrochloride |
Specifications
| 184°C to 190°C | |
| Purple to Red | |
| 98% | |
| Glass Bottle | |
| MFCD00067560 | |
| l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl | |
| C(C(C(=O)N)N)O.Cl | |
| +14.5° (20°C c=1,H2O) | |
| 140.567 | |
| 140.57 | |
| Crystalline Powder or Crystals |
| 65414-74-6 | |
| Authentic | |
| C3H9ClN2O2 | |
| HOCHCH(NH)CONH·HCl | |
| +13.5° to +15.5° (20°C, 589nm) (c=1, H2O) | |
| VURWDDZIWBGXCK-DKWTVANSSA-N | |
| (2S)-2-amino-3-hydroxypropanamide;hydrochloride | |
| 1 g | |
| 16212403 | |
| ≥97.5% (Argentometry) | |
| L-Serinamide hydrochloride |
RUO – Research Use Only