missing translation for 'onlineSavingsMsg'
Learn More

L-Serinamide hydrochloride, 98%, Thermo Scientific™

Catalog No. AC303160010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303160010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC303160010 Supplier Thermo Scientific Chemicals Supplier No. 303160010

Chemical Identifiers

CAS 65414-74-6
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00067560
InChI Key VURWDDZIWBGXCK-DKWTVANSSA-N
Synonym l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl
PubChem CID 16212403
IUPAC Name (2S)-2-amino-3-hydroxypropanamide;hydrochloride
SMILES C(C(C(=O)N)N)O.Cl

Specifications

Melting Point 184°C to 190°C
Color Purple to Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCHCH(NH)CONH·HCl
Specific Rotation +13.5° to +15.5° (20°C, 589nm) (c=1, H2O)
Specific Rotation Condition +14.5° (20°C c=1,H2O)
Quantity 1 g
Formula Weight 140.57
Percent Purity ≥97.5% (Argentometry)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Serinamide hydrochloride
Show More Show Less

RUO – Research Use Only