missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Serinamide hydrochloride, 95%
CAS: 65414-74-6 | C3H9ClN2O2 | 140.567 g/mol
$144.75 - $605.49
Chemical Identifiers
| CAS | 65414-74-6 |
|---|---|
| Molecular Formula | C3H9ClN2O2 |
| Molecular Weight (g/mol) | 140.567 |
| MDL Number | MFCD00067560 |
| InChI Key | VURWDDZIWBGXCK-DKWTVANSSA-N |
| Synonym | l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl |
| PubChem CID | 16212403 |
| IUPAC Name | (2S)-2-amino-3-hydroxypropanamide;hydrochloride |
| SMILES | C(C(C(=O)N)N)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6516906
|
Thermo Scientific Chemicals
H6516906 |
5 g |
Each for $144.75
|
|
|||||
|
AAH6516914
|
Thermo Scientific Chemicals
H6516914 |
25 g |
Each for $605.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 65414-74-6 | |
| 140.567 | |
| VURWDDZIWBGXCK-DKWTVANSSA-N | |
| 16212403 | |
| C(C(C(=O)N)N)O.Cl |
| C3H9ClN2O2 | |
| MFCD00067560 | |
| l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl | |
| (2S)-2-amino-3-hydroxypropanamide;hydrochloride |
Specifications
| 185°C to 188°C | |
| White | |
| MFCD00067560 | |
| l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl | |
| C(C(C(=O)N)N)O.Cl | |
| 5 g | |
| 16212403 | |
| 95% | |
| L-Serinamide hydrochloride |
| 65414-74-6 | |
| C3H9ClN2O2 | |
| 3911773 | |
| VURWDDZIWBGXCK-DKWTVANSSA-N | |
| (2S)-2-amino-3-hydroxypropanamide;hydrochloride | |
| 140.567 | |
| 140.57 | |
| Powder |
Safety and Handling
EINECSNumber : 265-753-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only