missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ L-Selenomethionine

Catalog No. A39247
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39247 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39247 Supplier Gibco™ Supplier No. A39247
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Selenomethionine is a naturally occurring amino acid containing selenium that is commonly used for the labeling of proteins to facilitate structural determination by x-ray crystallography.

Selenomethionine is an antioxidant and has been shown to increase glutathione peroxidase activity in endothelial cells

Specifications

Chemical Name or Material (S)-2-Amino-4-(methylseleno)butyric acid
Molecular Formula C5H11NO2Se
MDL Number MFCD00037210
InChI Key RJFAYQIBOAGBLC-BYPYZUCNSA-N
SMILES C[Se]CC[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-(methylselanyl)butanoic acid
Molecular Weight (g/mol) 196.12
Quantity 250 mg
Percent Purity >98%
Physical Form Powder
Shipping Condition Wet Ice
Show More Show Less
I am performing L-selenomethionine labeling and getting very low protein yields. What could be going wrong?

L-selenomethionine is a known cell toxin. Therefore, it is important to grow cells in the present of this amino acid for the shortest amount of time required to achieve maximal labeling. We recommend adding L-selenomethionine to the culture just prior to transfection (in Expi293 Met (-) Expression Medium). You may also starve cells for 6 hr by culturing cells in Expi293 Met (-) Expression Medium (without supplementation) and then proceed with L-selenomethionine add-back and transfection.

For Research Use Only. Not for use in diagnostic procedures.