Learn More
L-Selenocystine, 95%, Thermo Scientific Chemicals
CAS: 29621-88-3 | C6H12N2O4Se2 | 334.11 g/mol
Supplier: Thermo Scientific Chemicals 301862500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L-Selenocystine | |
| 29621-88-3 | |
| Yellow | |
| 98% | |
| C6H12N2O4Se2 | |
| 250 mg | |
| Solubility in water: soluble in water | |
| N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O | |
| 334.11 | |
| 334.09 | |
| Powder |
| 98% | |
| 212°C to 215°C | |
| Authentic | |
| Glass bottle | |
| MFCD00800971 | |
| l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer | |
| JULROCUWKLNBSN-IMJSIDKUSA-N | |
| (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid | |
| 207306 | |
| 98% |
Chemical Identifiers
| 29621-88-3 | |
| 334.11 | |
| JULROCUWKLNBSN-IMJSIDKUSA-N | |
| 207306 | |
| N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O |
| C6H12N2O4Se2 | |
| MFCD00800971 | |
| l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer | |
| (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RTECSNumber : AY6032000
RUO – Research Use Only