missing translation for 'onlineSavingsMsg'
Learn More

L-Selenocystine, 95%, Thermo Scientific Chemicals

CAS: 29621-88-3 | C6H12N2O4Se2 | 334.11 g/mol

Supplier:  Thermo Scientific Chemicals 301860010

 View more versions of this product

Catalog No. AC301860010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Selenocystine
29621-88-3
Yellow
98%
C6H12N2O4Se2
1 g
Solubility in water: soluble in water
N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O
334.11
334.09
Powder
98%
212°C to 215°C
Authentic
Glass bottle
MFCD00800971
l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer
JULROCUWKLNBSN-IMJSIDKUSA-N
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid
207306
98%

Chemical Identifiers

29621-88-3
334.11
JULROCUWKLNBSN-IMJSIDKUSA-N
207306
N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O
C6H12N2O4Se2
MFCD00800971
l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : AY6032000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only