missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ribose, 99%, Thermo Scientific™

Chemical Identifiers

CAS 24259-59-4
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00167010
InChI Key PYMYPHUHKUWMLA-MROZADKFSA-N
Synonym l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PubChem CID 90428
ChEBI CHEBI:47015
IUPAC Name (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@H](O)[C@H](O)[C@H](O)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC257910010
View Documents
Thermo Scientific Chemicals
257910010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC257910050
View Documents
Thermo Scientific Chemicals
257910050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC257910250
View Documents
Thermo Scientific Chemicals
257910250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

24259-59-4
150.13
PYMYPHUHKUWMLA-MROZADKFSA-N
90428
(2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00167010
l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
CHEBI:47015
OC[C@H](O)[C@H](O)[C@H](O)C=O

Specifications

24259-59-4
98.5% min. (ELSD) (HPLC)
C5H10O5
1g
+ 20.00 (20.00°C c=2,water)
l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PYMYPHUHKUWMLA-MROZADKFSA-N
(2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
90428
150.13
L(+)-Ribose
Authentic
Glass Bottle
MFCD00167010
01,859
+ 20.00
(10% in water) Clear colorless
OC[C@H](O)[C@H](O)[C@H](O)C=O
150.13
CHEBI:47015
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 246-110-4

SDS
Documents

Documents

RUO – Research Use Only