missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Rhamnose Monohydrate, Promega™
Description
- Induces Expression of ORFs in Vectors Transformed into Single Step (KRX) E. coli Cells
- Used with Single Step (KRX) Competent Cells
- Provides dramatic control of expressed protein-coding regions with higher protein yields
- Induces expression of T7 RNA Polymerase under control of rhamnose promoter rhaBAD
Specifications
Specifications
| CAS | 10030-85-0 |
| Quantity | 50 g |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Rhamnose Monohydrate |
Safety and Handling
| Recommended Storage | 15 to 30°C |