missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Pyroglutamic Acid 97.0+%, TCI America™
Supplier: TCI America P0573100G
Specifications
| L-Pyroglutamic Acid | |
| 160°C | |
| C5H7NO3 | |
| 100 g | |
| ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
| 5-oxopyrrolidine-2-carboxylic acid | |
| 7405 | |
| 129.12 | |
| Crystalline Powder |
| 98-79-3 | |
| White | |
| MFCD00005272 | |
| l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo | |
| OC(=O)C1CCC(=O)N1 | |
| 129.12 | |
| CHEBI:18183 | |
| ≥97.0% (T) |
Chemical Identifiers
| 98-79-3 | |
| 129.12 | |
| ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
| 7405 | |
| 5-oxopyrrolidine-2-carboxylic acid |
| C5H7NO3 | |
| MFCD00005272 | |
| l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo | |
| CHEBI:18183 | |
| OC(=O)C1CCC(=O)N1 |
Safety and Handling
EINECSNumber : (5)-0115
RTECSNumber : TW3710000
TSCA : Yes