missing translation for 'onlineSavingsMsg'
Learn More

L-Pyroglutamic Acid 97.0+%, TCI America™

Catalog No. P057325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P057325G 25 g
P0573100G 100 g
P0573500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P057325G Supplier TCI America Supplier No. P057325G

Chemical Identifiers

CAS 98-79-3
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005272
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo
PubChem CID 7405
ChEBI CHEBI:18183
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 129.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Pyroglutamic Acid
EINECSNumber (5)-0115
RTECSNumber TW3710000
TSCA Yes