missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Prolinol, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 418320050

 View more versions of this product

Catalog No. AC418320050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Authentic
23356-96-9
1.4840 to 1.4880
1.0360g/mL
5g
95%
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
[(2S)-pyrrolidin-2-yl]methanol
640091
95%
L(+)-Prolinol
Glass bottle
+ 31.00
+31° (20° (c=1,toluene))
86°C
C5H11NO
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
C1CC(NC1)CO
101.149
101.15

Chemical Identifiers

23356-96-9
101.149
HVVNJUAVDAZWCB-YFKPBYRVSA-N
640091
C1CC(NC1)CO
C5H11NO
MFCD00005255
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
[(2S)-pyrrolidin-2-yl]methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only