missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester, 98%

Catalog No. AAH6365706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6365706 5 g
AAH6365703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6365706 Supplier Thermo Scientific Chemicals Supplier No. H6365706
Only null left

CAS: 2812-46-6 | C9H17NO2 | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2812-46-6
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00037879
InChI Key XJJBXZIKXFOMLP-ZETCQYMHSA-N
Synonym l-proline t-butyl ester, h-pro-otbu, tert-butyl l-prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, l-proline tert-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, l-proline t-butyl ester free base, t-butyl prolinate
PubChem CID 6453108
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate
SMILES CC(C)(C)OC(=O)C1CCCN1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 171.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material L-Proline tert-butyl ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-558-0
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only