missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| L-Proline | |
| 228°C | |
| C5H9NO2 | |
| 250 g | |
| ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
| pyrrolidine-2-carboxylic acid | |
| 145742 | |
| 115.13 | |
| Crystalline Powder |
| 147-85-3 | |
| White | |
| MFCD00064318 | |
| l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
| OC(=O)C1CCCN1 | |
| 115.13 | |
| CHEBI:17203 | |
| ≥99.0% (HPLC,T) |
Chemical Identifiers
| 147-85-3 | |
| 115.13 | |
| ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
| 145742 | |
| pyrrolidine-2-carboxylic acid |
| C5H9NO2 | |
| MFCD00064318 | |
| l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
| CHEBI:17203 | |
| OC(=O)C1CCCN1 |
Safety and Handling
EINECSNumber : (9)-1626
RTECSNumber : TW3584000
TSCA : Yes