missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603975
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603975 25 g Poly Bottle
18603976 100 g Poly Bottle
18603977 1 kg Poly Pail
18603978 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603975 Supplier Spectrum Chemical Mfg Cor Supplier No. P143625GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1436, 147-85-3, C5H9NO2

Spectrum™ Chemical L-Proline, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Packaging Poly Bottle
Quantity 25 g
Optical Rotation -84.0° to -86.3°
Percent Purity 98.5 to 101.5%
Grade FCC