missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, >99%, MP Biomedicals™

Catalog No. ICN10273080
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10273080 100 g
ICN10273091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10273080 Supplier MP Biomedicals Inc Supplier No. 0210273080

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

Color White
Assay Percent Range >99%
Quantity 100 g
Formula Weight 115.13
Percent Purity >99%
Physical Form Powder
Chemical Name or Material L-Proline, Cell Biology and Analysis
EINECSNumber 205-702-2
Recommended Storage Store at Room Temperature.