missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, 98.5%, Spectrum™ Chemical

Catalog No. 18611383
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611383 25 g Poly Bottle
18611382 100 g Poly Bottle
18611381 1 kg Poly Pail
18611384 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611383 Supplier Spectrum Chemical Mfg Cor Supplier No. PR12025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 147-85-3

Specifications

CAS Max % 1
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 25 g
Specific Rotation -84.0° to -86.3°
Percent Purity 0.985
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.