missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Pipecolinic acid, 99%
Description
L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∽270°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 81093 |
| Merck Index | 14,7458 |
| Solubility Information | Soluble in water. |
| Optical Rotation | −26° (c=1 in Water) |
| Formula Weight | 129.16 |
| Percent Purity | 99% |
| Chemical Name or Material | L-Pipecolinic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 221-462-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only