missing translation for 'onlineSavingsMsg'
Learn More

L-Pipecolic Acid 98.0+%, TCI America™

Catalog No. p-7135119
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14041G 1 g
P14045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14041G Supplier TCI America Supplier No. P14041G

Chemical Identifiers

CAS 3105-95-1
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00005981
InChI Key HXEACLLIILLPRG-UHFFFAOYNA-N
Synonym l-pipecolic acid, s-piperidine-2-carboxylic acid, l-homoproline, h-hopro-oh, l-pipecolinic acid, l-pipecolate, 2s-piperidine-2-carboxylic acid, s-pipecolic acid, l--pipecolinic acid, l-piperidine-2-carboxylic acid
PubChem CID 439227
ChEBI CHEBI:30913
IUPAC Name piperidine-2-carboxylic acid
SMILES OC(=O)C1CCCCN1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Pipecolic Acid