missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-alpha-Phenylglycine, 99%

Catalog No. AC217991000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217991000 100 g Plastic bottle
AC217990250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC217991000 Supplier Thermo Scientific Chemicals Supplier No. 217991000
Only null left

CAS: 2935-35-5 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2935-35-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00064403
InChI Key ZGUNAGUHMKGQNY-ZETCQYMHSA-N
Synonym h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid
PubChem CID 99291
ChEBI CHEBI:439819
IUPAC Name (2S)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Melting Point >300.0°C
Color White
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH(NH2)CO2H
Quantity 100 g
Beilstein 14, 459
Fieser 13,225
Merck Index 14, 7291
Specific Rotation Condition + 157.00 (20.00°C c=2,2N HCl)
Specific Rotation + 157.00
Solubility Information Solubility in water: 0.3g/100mL. Other solubilities: soluble in aq.sol. of inorg. solvents and alkalies,practically insoluble in alcohol
Transmittance 98% min. (c=5, 2N HCl, 430nm) 1cm cell
Formula Weight 151.16
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-α-Phenylglycine, 99%
Show More Show Less
EINECSNumber 220-909-8
TSCA TSCA

RUO – Research Use Only