missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine tert-butyl ester hydrochloride, 98%

Catalog No. AAH5912206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5912206 5 g
AAH5912203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5912206 Supplier Thermo Scientific Chemicals Supplier No. H5912206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15100-75-1 | C13H20ClNO2 | 257.758 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 15100-75-1
Molecular Formula C13H20ClNO2
Molecular Weight (g/mol) 257.758
MDL Number MFCD00038894
InChI Key FDMCEXDXULPJPG-MERQFXBCSA-N
Synonym h-phe-otbu.hcl, h-phe-otbu hcl, tert-butyl 3-phenyl-l-alaninate hydrochloride, s-tert-butyl 2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine tert-butyl ester hydrochloride, l-phenylalanine t-butyl ester hydrochloride, tert-butyl 2s-2-amino-3-phenylpropanoate hydrochloride, phe-otbu.hcl, tert-butyl l-phenylalaninate hydrochloride, phenylalanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 11459563
IUPAC Name tert-butyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES CC(C)(C)OC(=O)C(CC1=CC=CC=C1)N.Cl

Specifications

Melting Point 240°C
Linear Formula C6H5CH2CH(NH2)COOC(CH3)3·HCl
Beilstein 4723424
Solubility Information Soluble in methanol.
Quantity 5 g
Formula Weight 257.76
Optical Rotation +47° (c=2 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Phenylalanine tert-butyl ester hydrochloride
EINECSNumber 239-151-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only